RIE156327111 Rieke Metals, Inc.

3-(4-chlorophenyl)-3',5'-difluoropropiophenone

  • MDL number MFCD03843497
  • Cas Number : 898788-49-3
  • Molecular Weight : 280.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-60
Availability details for
Quantity
  • Availability
  • Add to Cart