RIE156327112 Rieke Metals, Inc.

3-(4-chlorophenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843498
  • Cas Number : 898788-51-7
  • Molecular Weight : 298.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-61
Availability details for
Quantity
  • Availability
  • Add to Cart