RIE156327114 Rieke Metals, Inc.

2-(4-chlorophenyl)ethyl cyclopropyl ketone

  • MDL number MFCD03843499
  • Cas Number : 898788-55-1
  • Molecular Weight : 208.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2101-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2101-00
Availability details for
Quantity
  • Availability
  • Add to Cart