RIE156327115 Rieke Metals, Inc.

2-(4-Chlorophenyl)ethyl cyclobutyl ketone

  • MDL number MFCD03843500
  • Cas Number : 898788-57-3
  • Molecular Weight : 222.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2102-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2102-00
Availability details for
Quantity
  • Availability
  • Add to Cart