RIE156327116 Rieke Metals, Inc.

2-(4-chlorophenyl)ethyl cyclopentyl ketone

  • MDL number MFCD03843501
  • Cas Number : 898788-59-5
  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-cyclopentyl-1-propanone
Inchi Code 1S/C14H17ClO/c15-13-8-5-11(6-9-13)7-10-14(16)12-3-1-2-4-12/h5-6,8-9,12H,1-4,7,10H2
Inchi Key JAZWZVIWCSORLF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2103-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2103-00
Availability details for
Quantity
  • Availability
  • Add to Cart