RIE156327117 Rieke Metals, Inc.

2-(4-Chlorophenyl)ethyl cyclohexyl ketone

  • MDL number MFCD03843502
  • Cas Number : 898788-61-9
  • Molecular Weight : 250.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2104-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2104-00
Availability details for
Quantity
  • Availability
  • Add to Cart