RIE156327119 Rieke Metals, Inc.

3-(3-fluorophenyl)-2'-methylpropiophenone

  • MDL number MFCD03843504
  • Cas Number : 898788-65-3
  • Molecular Weight : 242.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-fluorophenyl)-1-(2-methylphenyl)-1-propanone
Inchi Code 1S/C16H15FO/c1-12-5-2-3-8-15(12)16(18)10-9-13-6-4-7-14(17)11-13/h2-8,11H,9-10H2,1H3
Inchi Key IKLUUVRHSUQIKF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2200-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-02
Availability details for
Quantity
  • Availability
  • Add to Cart