RIE156327120 Rieke Metals, Inc.

3-(3-fluorophenyl)-3'-methylpropiophenone

  • MDL number MFCD03843505
  • Cas Number : 898788-67-5
  • Molecular Weight : 242.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-fluorophenyl)-1-(3-methylphenyl)-1-propanone
Inchi Code 1S/C16H15FO/c1-12-4-2-6-14(10-12)16(18)9-8-13-5-3-7-15(17)11-13/h2-7,10-11H,8-9H2,1H3
Inchi Key YNIAJPZEFXMTJK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2200-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-03
Availability details for
Quantity
  • Availability
  • Add to Cart