RIE156327124 Rieke Metals, Inc.

3-(3-fluorophenyl)-4'-methoxypropiophenone

  • MDL number MFCD03843509
  • Cas Number : 898788-76-6
  • Molecular Weight : 258.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-07
Availability details for
Quantity
  • Availability
  • Add to Cart