RIE156327126 Rieke Metals, Inc.

3'-cyano-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843511
  • Cas Number : 898788-82-4
  • Molecular Weight : 253.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[3-(3-fluorophenyl)propanoyl]benzonitrile
Inchi Code 1S/C16H12FNO/c17-15-6-2-3-12(10-15)7-8-16(19)14-5-1-4-13(9-14)11-18/h1-6,9-10H,7-8H2
Inchi Key USDFCGJFWAAGMP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2200-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-09
Availability details for
Quantity
  • Availability
  • Add to Cart