RIE156327129 Rieke Metals, Inc.

3'-carboethoxy-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843514
  • Cas Number : 898788-91-5
  • Molecular Weight : 300.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-12
Availability details for
Quantity
  • Availability
  • Add to Cart