RIE156327131 Rieke Metals, Inc.

3-(3-fluorophenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD03843516
  • Cas Number : 898788-97-1
  • Molecular Weight : 274.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-fluorophenyl)-1-[2-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C16H15FOS/c1-19-16-8-3-2-7-14(16)15(18)10-9-12-5-4-6-13(17)11-12/h2-8,11H,9-10H2,1H3
Inchi Key ZOLZLXYGICVQRU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2200-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-14
Availability details for
Quantity
  • Availability
  • Add to Cart