RIE156327139 Rieke Metals, Inc.

2',3'-dimethyl-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843524
  • Cas Number : 898789-20-3
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-26
Availability details for
Quantity
  • Availability
  • Add to Cart