RIE156327141 Rieke Metals, Inc.

2',5'-dimethyl-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843526
  • Cas Number : 898767-11-8
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-28
Availability details for
Quantity
  • Availability
  • Add to Cart