RIE156327142 Rieke Metals, Inc.

2',6'-dimethyl-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843527
  • Cas Number : 898767-14-1
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-29
Availability details for
Quantity
  • Availability
  • Add to Cart