RIE156327143 Rieke Metals, Inc.

3',4'-Dimethyl-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843528
  • Cas Number : 898767-17-4
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-30
Availability details for
Quantity
  • Availability
  • Add to Cart