RIE156327144 Rieke Metals, Inc.

3',5'-dimethyl-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843529
  • Cas Number : 898767-20-9
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-31
Availability details for
Quantity
  • Availability
  • Add to Cart