RIE156327147 Rieke Metals, Inc.

3'-Chloro-4'-fluoro-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843532
  • Cas Number : 898767-29-8
  • Molecular Weight : 280.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2200-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-34
Availability details for
Quantity
  • Availability
  • Add to Cart