RIE156327156 Rieke Metals, Inc.

4'-chloro-2'-fluoro-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843541
  • Cas Number : 898767-55-0
  • Molecular Weight : 280.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chloro-2-fluorophenyl)-3-(3-fluorophenyl)-1-propanone
Inchi Code 1S/C15H11ClF2O/c16-11-5-6-13(14(18)9-11)15(19)7-4-10-2-1-3-12(17)8-10/h1-3,5-6,8-9H,4,7H2
Inchi Key NHLBMNLVWXNJDI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2200-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-52
Availability details for
Quantity
  • Availability
  • Add to Cart