RIE156327157 Rieke Metals, Inc.

2',3'-dichloro-3-(3-fluorophenyl)propiophenone

  • MDL number MFCD03843542
  • Cas Number : 898767-57-2
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,3-dichlorophenyl)-3-(3-fluorophenyl)-1-propanone
Inchi Code 1S/C15H11Cl2FO/c16-13-6-2-5-12(15(13)17)14(19)8-7-10-3-1-4-11(18)9-10/h1-6,9H,7-8H2
Inchi Key YEFADJPFNYLAFO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2200-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2200-53
Availability details for
Quantity
  • Availability
  • Add to Cart