RIE156327176 Rieke Metals, Inc.

3-(4-fluorophenyl)-3'-methoxypropiophenone

  • MDL number MFCD03843560
  • Cas Number : 898767-93-6
  • Molecular Weight : 258.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-06
Availability details for
Quantity
  • Availability
  • Add to Cart