RIE156327186 Rieke Metals, Inc.

3'-chloro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843572
  • Cas Number : 898768-19-9
  • Molecular Weight : 262.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-20
Availability details for
Quantity
  • Availability
  • Add to Cart