RIE156327187 Rieke Metals, Inc.

4'-chloro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843573
  • Cas Number : 654673-24-2
  • Molecular Weight : 262.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chlorophenyl)-3-(4-fluorophenyl)-1-propanone
Inchi Code 1S/C15H12ClFO/c16-13-6-4-12(5-7-13)15(18)10-3-11-1-8-14(17)9-2-11/h1-2,4-9H,3,10H2
Inchi Key WYRFWQBCJMKAGJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2300-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-21
Availability details for
Quantity
  • Availability
  • Add to Cart