RIE156327188 Rieke Metals, Inc.

3'-fluoro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843574
  • Cas Number : 898768-22-4
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-22
Availability details for
Quantity
  • Availability
  • Add to Cart