RIE156327189 Rieke Metals, Inc.

4'-fluoro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843575
  • Cas Number : 104147-29-7
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-23
Availability details for
Quantity
  • Availability
  • Add to Cart