RIE156327193 Rieke Metals, Inc.

2',6'-dimethyl-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843579
  • Cas Number : 898768-34-8
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-29
Availability details for
Quantity
  • Availability
  • Add to Cart