RIE156327205 Rieke Metals, Inc.

2'-chloro-4'-fluoro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843591
  • Cas Number : 898768-62-2
  • Molecular Weight : 280.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloro-4-fluorophenyl)-3-(4-fluorophenyl)-1-propanone
Inchi Code 1S/C15H11ClF2O/c16-14-9-12(18)6-7-13(14)15(19)8-3-10-1-4-11(17)5-2-10/h1-2,4-7,9H,3,8H2
Inchi Key VMCODRPPOQEXJR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2300-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-50
Availability details for
Quantity
  • Availability
  • Add to Cart