RIE156327207 Rieke Metals, Inc.

3-(4-chlorophenyl)-4'-methoxypropiophenone

  • MDL number MFCD03843457
  • Cas Number : 111302-55-7
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H15ClO2/c1-19-15-9-5-13(6-10-15)16(18)11-4-12-2-7-14(17)8-3-12/h2-3,5-10H,4,11H2,1H3
Inchi Key ZBTZYDKQZYEMDA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-07
Availability details for
Quantity
  • Availability
  • Add to Cart