RIE156327208 Rieke Metals, Inc.

Cyclohexyl-2-phenethyl ketone

  • MDL number MFCD00965990
  • Cas Number : 43125-06-0
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0104-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0104-00
Availability details for
Quantity
  • Availability
  • Add to Cart