RIE156327209 Rieke Metals, Inc.

3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842961
  • Cas Number : 84839-91-8
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-methylphenyl)-1-phenyl-1-propanone
Inchi Code 1S/C16H16O/c1-13-7-5-6-8-14(13)11-12-16(17)15-9-3-2-4-10-15/h2-10H,11-12H2,1H3
Inchi Key NLXWIEOJEBAXRH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-01
Availability details for
Quantity
  • Availability
  • Add to Cart