RIE156327211 Rieke Metals, Inc.

4'-Methoxy-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD00757267
  • Cas Number : 20615-47-8
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-07
Availability details for
Quantity
  • Availability
  • Add to Cart