RIE156327218 Rieke Metals, Inc.

3-(3-chlorophenyl)propiophenone

  • MDL number MFCD02065472
  • Cas Number : 58122-03-5
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-01
Availability details for
Quantity
  • Availability
  • Add to Cart