RIE156327220 Rieke Metals, Inc.

4'-chloro-3-(4-chlorophenyl)propiophenone

  • MDL number MFCD03843469
  • Cas Number : 97009-36-4
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-bis(4-chlorophenyl)-1-propanone
Inchi Code 1S/C15H12Cl2O/c16-13-6-1-11(2-7-13)3-10-15(18)12-4-8-14(17)9-5-12/h1-2,4-9H,3,10H2
Inchi Key ZDRPIYBPFYVFLT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-21
Availability details for
Quantity
  • Availability
  • Add to Cart