RIE156327223 Rieke Metals, Inc.

2',5'-dimethyl-3-phenylpropiophenone

  • MDL number MFCD03842933
  • Cas Number : 108976-70-1
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-28
Availability details for
Quantity
  • Availability
  • Add to Cart