RIE156327227 Rieke Metals, Inc.

4'-chloro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843163
  • Cas Number : 111302-58-0
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-21
Availability details for
Quantity
  • Availability
  • Add to Cart