RIE156327230 Rieke Metals, Inc.

cyclohexyl 2-(3-methoxyphenyl)ethyl ketone

  • MDL number MFCD03843146
  • Cas Number : 175234-19-2
  • Molecular Weight : 246.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclohexyl-3-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H22O2/c1-18-15-9-5-6-13(12-15)10-11-16(17)14-7-3-2-4-8-14/h5-6,9,12,14H,2-4,7-8,10-11H2,1H3
Inchi Key IBCODFIANGWDCZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0604-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0604-00
Availability details for
Quantity
  • Availability
  • Add to Cart