RIE156327235 Rieke Metals, Inc.

4'-Bromo-3-phenylpropiophenone

  • MDL number MFCD01851729
  • Cas Number : 1669-51-8
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-3-phenyl-1-propanone
Inchi Code 1S/C15H13BrO/c16-14-9-7-13(8-10-14)15(17)11-6-12-4-2-1-3-5-12/h1-5,7-10H,6,11H2
Inchi Key AKQSTOHWSLTIIU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0100-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-19
Availability details for
Quantity
  • Availability
  • Add to Cart