RIE156328046 Rieke Metals, Inc.

2',4'-dichloro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843595
  • Cas Number : 898768-70-2
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-54
Availability details for
Quantity
  • Availability
  • Add to Cart