RIE156328049 Rieke Metals, Inc.

3',5'-dichloro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843598
  • Cas Number : 898768-76-8
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-57
Availability details for
Quantity
  • Availability
  • Add to Cart