RIE156328050 Rieke Metals, Inc.

2',4'-Difluoro-3-(4-fluorophenyl)propiophenone

  • MDL number MFCD03843599
  • Cas Number : 654673-34-4
  • Molecular Weight : 264.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2300-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2300-58
Availability details for
Quantity
  • Availability
  • Add to Cart