RIE156328055 Rieke Metals, Inc.

cyclopropyl 2-(4-fluorophenyl)ethyl ketone

  • MDL number MFCD03843603
  • Cas Number : 898768-86-0
  • Molecular Weight : 192.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropyl-3-(4-fluorophenyl)-1-propanone
Inchi Code 1S/C12H13FO/c13-11-6-1-9(2-7-11)3-8-12(14)10-4-5-10/h1-2,6-7,10H,3-5,8H2
Inchi Key IROFXNWHNPHFNS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2301-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2301-00
Availability details for
Quantity
  • Availability
  • Add to Cart