RIE156328056 Rieke Metals, Inc.

Cyclobutyl 2-(4-fluorophenyl)ethyl ketone

  • MDL number MFCD03843604
  • Cas Number : 898768-88-2
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2302-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2302-00
Availability details for
Quantity
  • Availability
  • Add to Cart