RIE156328057 Rieke Metals, Inc.

cyclopentyl 2-(4-fluorophenyl)ethyl ketone

  • MDL number MFCD03843605
  • Cas Number : 898768-90-6
  • Molecular Weight : 220.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2303-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2303-00
Availability details for
Quantity
  • Availability
  • Add to Cart