RIE156328058 Rieke Metals, Inc.

cyclohexyl 2-(4-fluorophenyl)ethyl ketone

  • MDL number MFCD03843606
  • Cas Number : 898768-92-8
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2304-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2304-00
Availability details for
Quantity
  • Availability
  • Add to Cart