RIE156328061 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-3'-methylpropiophenone

  • MDL number MFCD03843609
  • Cas Number : 898768-98-4
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-03
Availability details for
Quantity
  • Availability
  • Add to Cart