RIE156328062 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-4'-methylpropiophenone

  • MDL number MFCD03843610
  • Cas Number : 898769-00-1
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2,3-dimethylphenyl)-1-(4-methylphenyl)-1-propanone
Inchi Code 1S/C18H20O/c1-13-7-9-17(10-8-13)18(19)12-11-16-6-4-5-14(2)15(16)3/h4-10H,11-12H2,1-3H3
Inchi Key ZENNHPKZHIDDKD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-04
Availability details for
Quantity
  • Availability
  • Add to Cart