RIE156328064 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-3'-methoxypropiophenone

  • MDL number MFCD03843612
  • Cas Number : 898769-05-6
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2,3-dimethylphenyl)-1-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C18H20O2/c1-13-6-4-7-15(14(13)2)10-11-18(19)16-8-5-9-17(12-16)20-3/h4-9,12H,10-11H2,1-3H3
Inchi Key YUXVDSVAOPOBCY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-06
Availability details for
Quantity
  • Availability
  • Add to Cart