RIE156328065 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-4'-methoxypropiophenone

  • MDL number MFCD03843613
  • Cas Number : 898769-08-9
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-07
Availability details for
Quantity
  • Availability
  • Add to Cart