RIE156328074 Rieke Metals, Inc.

3'-bromo-3-(2,3-dimethylphenyl)propiophenone

  • MDL number MFCD03843622
  • Cas Number : 898769-35-2
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-18
Availability details for
Quantity
  • Availability
  • Add to Cart