RIE156328075 Rieke Metals, Inc.

4'-bromo-3-(2,3-dimethylphenyl)propiophenone

  • MDL number MFCD03843623
  • Cas Number : 898769-38-5
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-19
Availability details for
Quantity
  • Availability
  • Add to Cart